Assay Details
Gene Symbol: AT4G16270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117724.4 | NP_193362.3 | 3 - 4 | 648 | 65 | |
GenBank mRNA | BT011747.1 | - | 3 - 4 | 648 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
