Assay Details
Gene Symbol: AT4G16555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341115.1 | NP_001328470.1 | 1 - 2 | 119 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
