Assay Details
Gene Symbol: MES16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117770.5 | NP_193402.1 | 2 - 3 | 736 | 93 | |
GenBank mRNA | AY088316.1 | - | 2 - 3 | 555 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
