Assay Details
Gene Symbol: MYB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341242.1 | NP_001329227.1 | 1 - 2 | 325 | 91 | |
NM_117888.3 | NP_567540.2 | 1 - 2 | 325 | 91 | ||
GenBank mRNA | AF175989.1 | - | 1 - 2 | 261 | 91 | |
AK118728.1 | - | 1 - 2 | 325 | 91 | ||
AY550297.1 | - | 1 - 2 | 261 | 91 | ||
BT005642.1 | - | 1 - 2 | 261 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
