Assay Details
Gene Symbol: GOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341276.1 | NP_001329915.1 | 10 - 11 | 1156 | 70 | |
NM_001341277.1 | NP_001329914.1 | 11 - 12 | 1097 | 70 | ||
NM_117946.3 | NP_193570.1 | 11 - 12 | 1053 | 70 | ||
GenBank mRNA | BT001945.1 | - | 10 - 11 | 922 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
