Assay Details
Gene Symbol: GOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084937.1 | NP_001078406.1 | 2 - 3 | 191 | 105 | |
NM_001341276.1 | NP_001329915.1 | 2 - 3 | 273 | 105 | ||
NM_001341277.1 | NP_001329914.1 | 2 - 3 | 315 | 105 | ||
NM_117946.3 | NP_193570.1 | 2 - 3 | 271 | 105 | ||
GenBank mRNA | BT001945.1 | - | 1 - 2 | 140 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
