Assay Details
Gene Symbol: GOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084937.1 | NP_001078406.1 | 8 - 9 | 687 | 82 | |
NM_001341276.1 | NP_001329915.1 | 8 - 9 | 769 | 82 | ||
NM_001341277.1 | NP_001329914.1 | 8 - 9 | 811 | 82 | ||
NM_117946.3 | NP_193570.1 | 8 - 9 | 767 | 82 | ||
GenBank mRNA | BT001945.1 | - | 7 - 8 | 636 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
