Assay Details
Gene Symbol: GOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084937.1 | NP_001078406.1 | 9 - 10 | 764 | 70 | |
NM_001341277.1 | NP_001329914.1 | 9 - 10 | 888 | 70 | ||
NM_117946.3 | NP_193570.1 | 9 - 10 | 844 | 70 | ||
GenBank mRNA | BT001945.1 | - | 8 - 9 | 713 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
