Assay Details
Gene Symbol: MYB98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117993.3 | NP_193612.1 | 1 - 2 | 713 | 129 | |
GenBank mRNA | AF176003.2 | - | 1 - 2 | 737 | 129 | |
DQ446849.1 | - | 1 - 2 | 675 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
