Assay Details
Gene Symbol: MYB98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117993.3 | NP_193612.1 | 2 - 3 | 836 | 68 | |
GenBank mRNA | AF176003.2 | - | 2 - 3 | 860 | 68 | |
DQ446849.1 | - | 2 - 3 | 798 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
