Assay Details
Gene Symbol: AT1G66850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105356.4 | NP_176857.1 | 1 | 418 | 135 | |
GenBank mRNA | AK117859.1 | - | 1 | 333 | 135 | |
AY086433.1 | - | 1 | 336 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
