Assay Details
Gene Symbol: AT1G68980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105570.3 | NP_177062.1 | 1 | 198 | 97 | |
GenBank mRNA | AK226281.1 | - | 1 | 155 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
