Assay Details
Gene Symbol: SIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105589.4 | NP_564955.1 | 13 - 14 | 1925 | 88 | |
NM_179534.2 | NP_849865.1 | 13 - 14 | 1730 | 88 | ||
GenBank mRNA | AY072199.1 | - | 13 - 14 | 1730 | 88 | |
AY133857.1 | - | 13 - 14 | 1678 | 88 | ||
U96613.1 | - | 13 - 14 | 1763 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
