Assay Details
Gene Symbol: SIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105589.4 | NP_564955.1 | 16 - 17 | 2497 | 79 | |
NM_179534.2 | NP_849865.1 | 16 - 17 | 2302 | 79 | ||
GenBank mRNA | AY072199.1 | - | 16 - 17 | 2302 | 79 | |
AY133857.1 | - | 16 - 17 | 2250 | 79 | ||
U96613.1 | - | 16 - 17 | 2335 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
