Assay Details
Gene Symbol: AT1G69350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105602.3 | NP_564961.1 | 1 | 2267 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AT1G69350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105602.3 | NP_564961.1 | 1 | 2267 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |