Assay Details
Gene Symbol: AT1G71695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105826.4 | NP_177313.1 | 1 - 2 | 536 | 72 | |
GenBank mRNA | AF334732.1 | - | 1 - 2 | 264 | 72 | |
BT000715.1 | - | 1 - 2 | 315 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
