Assay Details
Gene Symbol: AT1G71695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105826.4 | NP_177313.1 | 2 - 3 | 730 | 87 | |
GenBank mRNA | AF334732.1 | - | 2 - 3 | 458 | 87 | |
AY087964.1 | - | 2 - 3 | 512 | 87 | ||
BT000715.1 | - | 2 - 3 | 509 | 87 | ||
X98773.1 | - | 490 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
