Assay Details
Gene Symbol: EDA30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337489.1 | NP_001325638.1 | 6 - 7 | 1274 | 71 | |
NM_111252.5 | NP_187031.2 | 7 - 8 | 1186 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
