Assay Details
Gene Symbol: CHR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339086.1 | NP_001327912.1 | 4 - 5 | 1941 | 92 | |
NM_001339087.1 | NP_001327910.1 | 4 - 5 | 1832 | 92 | ||
NM_114134.2 | NP_189853.1 | 3 - 4 | 1694 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
