Assay Details
Gene Symbol: CHR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339086.1 | NP_001327912.1 | 5 - 6 | 2084 | 81 | |
NM_001339087.1 | NP_001327910.1 | 5 - 6 | 1975 | 81 | ||
NM_001339088.1 | NP_001327911.1 | 4 - 5 | 1843 | 81 | ||
NM_114134.2 | NP_189853.1 | 4 - 5 | 1837 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
