Assay Details
Gene Symbol: CR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126439.4 | NP_178487.1 | 13 - 14 | 1658 | 126 | |
GenBank mRNA | AF436826.1 | - | 13 - 14 | 1512 | 126 | |
AX765864.1 | - | 13 - 14 | 1429 | 126 | ||
AY053403.1 | - | 13 - 14 | 1497 | 126 | ||
BT002234.1 | - | 13 - 14 | 1429 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
