Assay Details
Gene Symbol: CR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126439.4 | NP_178487.1 | 5 - 6 | 857 | 93 | |
NM_179601.1 | NP_849932.1 | 5 - 6 | 711 | 93 | ||
GenBank mRNA | AF436826.1 | - | 5 - 6 | 711 | 93 | |
AX765864.1 | - | 5 - 6 | 628 | 93 | ||
AY053403.1 | - | 5 - 6 | 696 | 93 | ||
BT002234.1 | - | 5 - 6 | 628 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
