Assay Details
Gene Symbol: RPA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126648.4 | NP_178690.1 | 2 - 3 | 413 | 151 | |
NM_201704.2 | NP_973433.1 | 3 - 4 | 271 | 151 | ||
GenBank mRNA | AK118699.1 | - | 2 - 3 | 377 | 151 | |
BT005946.1 | - | 1 - 2 | 270 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
