Assay Details
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335345.1 | NP_001323939.1 | 23 - 24 | 2674 | 103 | |
NM_001335346.1 | NP_001323940.1 | 23 - 24 | 2674 | 103 | ||
NM_001335347.1 | NP_001323936.1 | 22 - 23 | 2591 | 103 | ||
NM_001335348.1 | NP_001323937.1 | 22 - 23 | 2520 | 103 | ||
NM_001335349.1 | NP_001323938.1 | 20 - 21 | 2451 | 103 | ||
NM_126770.8 | NP_178811.7 | 22 - 23 | 2531 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
