Assay Details
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335345.1 | NP_001323939.1 | 28 - 29 | 3297 | 93 | |
NM_001335346.1 | NP_001323940.1 | 28 - 29 | 3297 | 93 | ||
NM_001335347.1 | NP_001323936.1 | 27 - 28 | 3214 | 93 | ||
NM_001335348.1 | NP_001323937.1 | 27 - 28 | 3143 | 93 | ||
NM_001335349.1 | NP_001323938.1 | 25 - 26 | 3074 | 93 | ||
NM_126770.8 | NP_178811.7 | 27 - 28 | 3154 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
