Assay Details
Gene Symbol: NUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126895.4 | NP_178939.2 | 2 - 3 | 498 | 138 | |
GenBank mRNA | BT022114.1 | - | 2 - 3 | 426 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
