Assay Details
Gene Symbol: NUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126895.4 | NP_178939.2 | 6 - 7 | 920 | 64 | |
GenBank mRNA | BT022114.1 | - | 6 - 7 | 848 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
