Assay Details
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337542.1 | NP_001327439.1 | 11 - 12 | 1292 | 84 | |
NM_111353.4 | NP_187132.1 | 12 - 13 | 940 | 84 | ||
NM_202492.3 | NP_974221.2 | 11 - 12 | 932 | 84 | ||
GenBank mRNA | BT000875.1 | - | 10 - 11 | 618 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
