Assay Details
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337542.1 | NP_001327439.1 | 6 - 7 | 981 | 90 | |
NM_111353.4 | NP_187132.1 | 7 - 8 | 629 | 90 | ||
NM_202492.3 | NP_974221.2 | 6 - 7 | 621 | 90 | ||
GenBank mRNA | BT000875.1 | - | 5 - 6 | 307 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
