Assay Details
Gene Symbol: SHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118552.4 | NP_194150.1 | 6 - 7 | 859 | 103 | |
NM_202877.1 | NP_974606.1 | 6 - 7 | 706 | 103 | ||
GenBank mRNA | AB064528.1 | - | 6 - 7 | 692 | 103 | |
AY039895.1 | - | 6 - 7 | 700 | 103 | ||
BT004527.1 | - | 6 - 7 | 652 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
