Assay Details
Gene Symbol: AT4G24970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118629.3 | NP_194227.2 | 4 - 5 | 873 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
