Assay Details
Gene Symbol: AT4G28780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119022.3 | NP_194607.1 | 1 - 2 | 380 | 77 | |
GenBank mRNA | AY087166.1 | - | 1 - 2 | 308 | 77 | |
BT002993.1 | - | 1 - 2 | 308 | 77 | ||
BT004465.1 | - | 1 - 2 | 250 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
