Assay Details
Gene Symbol: AT4G28780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119022.3 | NP_194607.1 | 2 - 3 | 506 | 81 | |
GenBank mRNA | AY087166.1 | - | 2 - 3 | 434 | 81 | |
BT002993.1 | - | 2 - 3 | 434 | 81 | ||
BT004465.1 | - | 2 - 3 | 376 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
