Assay Details
Gene Symbol: PFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119080.4 | NP_194664.1 | 2 - 3 | 509 | 102 | |
GenBank mRNA | AK319124.1 | - | 3 - 4 | 331 | 102 | |
AY074350.1 | - | 2 - 3 | 355 | 102 | ||
AY114048.1 | - | 2 - 3 | 262 | 102 | ||
U43326.1 | - | 2 - 3 | 305 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
