Assay Details
Gene Symbol: AT4G29400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119085.4 | NP_194669.2 | 3 - 4 | 633 | 70 | |
GenBank mRNA | AY735656.1 | - | 3 - 4 | 602 | 70 | |
AY924824.1 | - | 3 - 4 | 550 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
