Assay Details
Gene Symbol: AT4G29400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119085.4 | NP_194669.2 | 4 - 5 | 687 | 63 | |
GenBank mRNA | AY735656.1 | - | 4 - 5 | 656 | 63 | |
AY924824.1 | - | 4 - 5 | 604 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
