Assay Details
Gene Symbol: AT4G30170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119163.3 | NP_194746.1 | 1 - 2 | 317 | 80 | |
GenBank mRNA | AF370284.1 | - | 1 - 2 | 238 | 80 | |
AY063051.1 | - | 1 - 2 | 214 | 80 | ||
AY085450.1 | - | 1 - 2 | 295 | 80 | ||
X98855.1 | - | 227 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
