Assay Details
Gene Symbol: AT4G30170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119163.3 | NP_194746.1 | 3 - 4 | 678 | 114 | |
GenBank mRNA | AF370284.1 | - | 3 - 4 | 599 | 114 | |
AY063051.1 | - | 3 - 4 | 575 | 114 | ||
AY085450.1 | - | 3 - 4 | 656 | 114 | ||
X98855.1 | - | 588 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
