Assay Details
Gene Symbol: ATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119204.4 | NP_194787.2 | 6 - 7 | 906 | 75 | |
GenBank mRNA | AK118028.1 | - | 6 - 7 | 897 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
