Assay Details
Gene Symbol: MYB49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124803.3 | NP_200234.1 | 2 - 3 | 480 | 137 | |
GenBank mRNA | AF175991.1 | - | 2 - 3 | 264 | 137 | |
AY059820.1 | - | 2 - 3 | 352 | 137 | ||
AY081512.1 | - | 2 - 3 | 264 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
