Assay Details
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345118.1 | NP_001330303.1 | 1 - 2 | 210 | 116 | |
NM_124899.4 | NP_200328.1 | 1 - 2 | 210 | 116 | ||
GenBank mRNA | AF510521.1 | - | 1 - 2 | 76 | 116 | |
BT024755.1 | - | 1 - 2 | 76 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
