Assay Details
Gene Symbol: AT5G55450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124927.4 | NP_568824.1 | 1 - 2 | 546 | 70 | |
GenBank mRNA | AY065290.1 | - | 1 - 2 | 393 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
