Assay Details
Gene Symbol: MYB80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124993.1 | NP_200422.1 | 1 - 2 | 137 | 62 | |
GenBank mRNA | AB493793.1 | - | 1 - 2 | 137 | 62 | |
AY519639.1 | - | 1 - 2 | 137 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
