Assay Details
Gene Symbol: LLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124999.5 | NP_200428.1 | 2 - 3 | 320 | 85 | |
GenBank mRNA | BT000437.1 | - | 2 - 3 | 250 | 85 | |
BT001157.1 | - | 2 - 3 | 147 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
