Assay Details
Gene Symbol: MGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342114.1 | NP_001329161.1 | 1 - 2 | 758 | 71 | |
NM_119327.3 | NP_194906.1 | 1 - 2 | 747 | 71 | ||
GenBank mRNA | AB047399.1 | - | 1 - 2 | 628 | 71 | |
AF241797.1 | - | 1 - 2 | 683 | 71 | ||
AY092965.1 | - | 1 - 2 | 824 | 71 | ||
BT008890.1 | - | 1 - 2 | 524 | 71 | ||
KJ138730.1 | - | 1 - 2 | 524 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
