Assay Details
Gene Symbol: MGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342113.1 | NP_001320109.1 | 4 - 5 | 1253 | 74 | |
NM_001342114.1 | NP_001329161.1 | 3 - 4 | 1318 | 74 | ||
NM_119327.3 | NP_194906.1 | 4 - 5 | 1229 | 74 | ||
GenBank mRNA | AB047399.1 | - | 4 - 5 | 1110 | 74 | |
AF241797.1 | - | 4 - 5 | 1165 | 74 | ||
AY092965.1 | - | 4 - 5 | 1306 | 74 | ||
BT008890.1 | - | 4 - 5 | 1006 | 74 | ||
KJ138730.1 | - | 4 - 5 | 1006 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
