Assay Details
Gene Symbol: MGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342113.1 | NP_001320109.1 | 6 - 7 | 1540 | 99 | |
NM_001342114.1 | NP_001329161.1 | 5 - 6 | 1605 | 99 | ||
NM_119327.3 | NP_194906.1 | 6 - 7 | 1516 | 99 | ||
GenBank mRNA | AB047399.1 | - | 6 - 7 | 1397 | 99 | |
AF241797.1 | - | 6 - 7 | 1452 | 99 | ||
AY092965.1 | - | 6 - 7 | 1593 | 99 | ||
BT008890.1 | - | 6 - 7 | 1293 | 99 | ||
KJ138730.1 | - | 6 - 7 | 1293 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
