Assay Details
Gene Symbol: PLDDELTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119745.3 | NP_567989.1 | 6 - 7 | 2101 | 112 | |
NM_179170.3 | NP_849501.1 | 6 - 7 | 2305 | 112 | ||
NM_179171.2 | NP_849502.1 | 6 - 7 | 2068 | 112 | ||
GenBank mRNA | AB031047.1 | - | 6 - 7 | 1915 | 112 | |
AF306345.1 | - | 6 - 7 | 1779 | 112 | ||
AF322228.1 | - | 6 - 7 | 1788 | 112 | ||
AK227213.1 | - | 6 - 7 | 1911 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
