Assay Details
Gene Symbol: PLDDELTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119745.3 | NP_567989.1 | 9 - 10 | 2605 | 65 | |
NM_179170.3 | NP_849501.1 | 9 - 10 | 2809 | 65 | ||
NM_179171.2 | NP_849502.1 | 8 - 9 | 2711 | 65 | ||
GenBank mRNA | AB031047.1 | - | 9 - 10 | 2419 | 65 | |
AF306345.1 | - | 9 - 10 | 2283 | 65 | ||
AF322228.1 | - | 9 - 10 | 2292 | 65 | ||
AK227213.1 | - | 9 - 10 | 2415 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
