Assay Details
Gene Symbol: KAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342508.1 | NP_001329354.1 | 14 - 15 | 5751 | 100 | |
NM_179236.3 | NP_849567.2 | 14 - 15 | 5751 | 100 | ||
GenBank mRNA | AY265959.1 | - | 14 - 15 | 5343 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
